Kallikrein inhibitor, solid

Supplier: Thermo Scientific Chemicals

J66234-LB0 J66234-MCR
AAJ66234-LB0EA 217.1 USD
AAJ66234-LB0 AAJ66234-MCR
Kallikrein inhibitor, solid
Kallikrein inhibitor
Formula: C₃₅H₅₅N₁₁O₉
MW: 773.89 g/mol
Storage Temperature: Freezer
CAS Number: 97145-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR