5-Bromo-2-fluoro-4-methylpyridine 96%

Warning

H61431-22 H61431-06 H61431-06 H61431-14 H61431-14 H61431-22
AAH61431-22EA 356.31 USD
AAH61431-22 CAAAH61431-06 AAH61431-14 AAH61431-06 CAAAH61431-14 CAAAH61431-22
5-Bromo-2-fluoro-4-methylpyridine 96%
5-Bromo-2-fluoro-4-methylpyridine
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839213
CAS Number: 864830-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR