4-Hydroxy-6-methylpyrimidine 97%

Danger

Synonyms: 6-Methylpyrimidin-4-ol

H61404-03 H61404-03
AAH61404-03EA 52.11 USD
AAH61404-03 CAAAH61404-03
4-Hydroxy-6-methylpyrimidine 97%
4-Hydroxy-6-methylpyrimidine
Formula: C₅H₆N₂O
MW: 110.116 g/mol
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00053575
CAS Number: 3524-87-6
EINECS: 222-543-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR