Methyl-2,6-difluoro-3-nitrobenzoate 98%

Warning

Synonyms: Methyl 2,6-difluoro-3-nitrobenzoate

H61327-03 H61327-03 H61327-06 H61327-14 H61327-14 H61327-06
AAH61327-03EA 24.98 USD
AAH61327-03 CAAAH61327-03 AAH61327-14 CAAAH61327-14 AAH61327-06 CAAAH61327-06
Methyl-2,6-difluoro-3-nitrobenzoate 98%
Methyl-2,6-difluoro-3-nitrobenzoate
Formula: C₈H₅F₂NO₄
MW: 217.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368850
CAS Number: 84832-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR