4-Benzyloxybenzotrifluoride 95%

Warning

H61303-06 H61303-03 H61303-MD H61303-03 H61303-MD H61303-06
AAH61303-03EA 44.95 USD
CAAAH61303-06 CAAAH61303-MD AAH61303-03 AAH61303-06 AAH61303-MD CAAAH61303-03
4-Benzyloxybenzotrifluoride 95%
4-Benzyloxybenzotrifluoride
Formula: C₁₄H₁₁F₃O
MW: 252.23
Storage Temperature: Ambient
MDL Number: MFCD00275905
CAS Number: 70097-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR