2-Amino-3-nitro-5-fluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-fluoro-3-nitropyridine 5-Fluoro-3-nitro-2-pyridylamine 5-Fluoro-3-nitropyridin-2-amine

H61293-06 H61293-03 H61293-03 H61293-06
AAH61293-03EA 120.8 USD
CAAAH61293-03 AAH61293-03 CAAAH61293-06 AAH61293-06
2-Amino-3-nitro-5-fluoropyridine 98%
2-Amino-3-nitro-5-fluoropyridine
Formula: C₅H₄FN₃O₂
MW: 157.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972413
CAS Number: 212268-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR