Azacyclonol 95%

Warning

Synonyms: alpha,alpha-Diphenyl-4-piperidinemethanolalpha-(4-Piperidyl)benzhydrolDiphenyl(4-piperidinyl)methanolDiphenyl-4-piperidinemethanolα,α-Diphenyl-4-piperidinomethanol

H61281-14 H61281-22 H61281-22 H61281-14
AAH61281-22EA 313.14 USD
AAH61281-22 CAAAH61281-14 CAAAH61281-22 AAH61281-14
Azacyclonol 95%
Azacyclonol
Formula: C₁₈H₂₁NO
MW: 267.37
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00066980
CAS Number: 115-46-8
EINECS: 204-092-5
Merck Index: 13,895

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR