2-Methoxy-4-methyl-5-nitropyridine 95%
|
Danger
|
H61258-06
H61258-03
H61258-03
H61258-06
AAH61258-03EA
40.01
USD
AAH61258-03
CAAAH61258-03
CAAAH61258-06
AAH61258-06
2-Methoxy-4-methyl-5-nitropyridine 95%
2-Methoxy-4-methyl-5-nitropyridine
Formula:
C₇H₈N₂O₃ MW: 168.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095075 CAS Number: 6635-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...