2-Amino-3-methylpyrazine 95%

Warning

Synonyms: 3-Methylpyrazin-2-amine

H61255-MD H61255-03 H61255-03 H61255-MD
AAH61255-MDEA 99.56 USD
CAAAH61255-03 CAAAH61255-MD AAH61255-MD AAH61255-03
2-Amino-3-methylpyrazine 95%
2-Amino-3-methylpyrazine
Formula: C₅H₇N₃
MW: 109.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705770
CAS Number: 19838-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR