4-Ethynylbenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H61244-06 H61244-03 H61244-06 H61244-03
AAH61244-06EA 313.99 USD
CAAAH61244-06 CAAAH61244-03 AAH61244-06 AAH61244-03
4-Ethynylbenzonitrile 97%
4-Ethynylbenzonitrile
Formula: C₉H₅N
MW: 127.14 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD04974058
CAS Number: 3032-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR