Methyl-3-fluoro-4-hydroxybenzoate 98%

Warning

Synonyms: Methyl 3-fluoro-4-hydroxybenzoate

H61219-06 H61219-MD H61219-MD H61219-03 H61219-03 H61219-06
AAH61219-06EA 279.13 USD
CAAAH61219-06 CAAAH61219-MD CAAAH61219-03 AAH61219-06 AAH61219-MD AAH61219-03
Methyl-3-fluoro-4-hydroxybenzoate 98%
Methyl 3-fluoro-4-hydroxybenzoate
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00215838
CAS Number: 403-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR