5-Fluoronicotinic acid 98%
|
Warning
|
H61162-22
H61162-14
H61162-06
H61162-06
H61162-14
H61162-22
AAH61162-14EA
152.52
USD
CAAAH61162-06
CAAAH61162-14
AAH61162-14
AAH61162-22
AAH61162-06
CAAAH61162-22
5-Fluoronicotinic acid 98%
5-Fluoronicotinic acid
Formula:
C₆H₄FNO₂ MW: 141.10 g/mol Melting Pt: 193…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318537 CAS Number: 402-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...