3-Fluoro-5-nitrotoluene 98%

Warning

H61100-MD H61100-03 H61100-06 H61100-06 H61100-MD H61100-03
AAH61100-MDEA 29.22 USD
CAAAH61100-03 CAAAH61100-06 AAH61100-MD AAH61100-06 CAAAH61100-MD AAH61100-03
3-Fluoro-5-nitrotoluene 98%
3-Fluoro-5-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797943
CAS Number: 499-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR