1,4-Bis(4-pyridylmethyl)benzene 97%

Warning

H37485-06 H37485-03 H37485-03 H37485-06
AAH37485-03EA 55.01 USD
CAAAH37485-03 CAAAH37485-06 AAH37485-03 AAH37485-06
1,4-Bis(4-pyridylmethyl)benzene 97%
1,4-Bis(4-pyridylmethyl)benzene
Formula: C₁₈H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD23379844
CAS Number: 52469-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR