2-Nitro-4-(trifluoromethyl)anisole 98%

Danger

Synonyms: Phosphorus (V) oxide chloride Phosphorus oxychloride

A11847-06 A11847-14 A11847-22 A11847-22 A11847-06 A11847-14
AAA11847-06EA 34.63 USD
AAA11847-06 CAAAA11847-14 CAAAA11847-06 CAAAA11847-22 AAA11847-22 AAA11847-14
2-Nitro-4-(trifluoromethyl)anisole 98%
2-Nitro-4-(trifluoromethyl)anisole
Formula: POCl₃
MW: 221.14 g/mol
Boiling Pt: 96…98 °C/0.6 mm
Melting Pt: 44…49 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007099
CAS Number: 394-25-2
EINECS: 206-891-4
UN: 1760
ADR: "8",III
Merck Index: 12,07506

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR