2-Amino-6-methoxypyrazine 95%

Warning

H61990-MD H61990-03 H61990-MD H61990-03
AAH61990-MDEA 100.67 USD
AAH61990-MD CAAAH61990-MD AAH61990-03 CAAAH61990-03
2-Amino-6-methoxypyrazine 95%
2-Amino-6-methoxypyrazine
Formula: C₅H₇N₃O
MW: 125.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909689
CAS Number: 6905-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR