4-(2-Fluoro-4-nitrophenyl)morpholine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61940-06 H61940-22 H61940-14 H61940-22 H61940-14 H61940-06
AAH61940-14EA 89.69 USD
AAH61940-14 CAAAH61940-14 AAH61940-22 CAAAH61940-06 CAAAH61940-22 AAH61940-06
4-(2-Fluoro-4-nitrophenyl)morpholine 98%
4-(2-Fluoro-4-nitrophenyl)morpholine
Formula: C₁₀H₁₁FN₂O₃
MW: 226.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03138381
CAS Number: 2689-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR