2-Amino-5-bromo-2'-fluorobenzophenone 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (2-Amino-5-bromophenyl)(2-fluorophenyl)methanone

H61882-06 H61882-06 H61882-03
AAH61882-06EA 124 USD
AAH61882-06 CAAAH61882-03 CAAAH61882-06
2-Amino-5-bromo-2'-fluorobenzophenone 95%
2-Amino-5-bromo-2'-fluorobenzophenone
Formula: C₁₃H₉BrFNO
MW: 294.12 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00038380
CAS Number: 1479-58-9
EINECS: 216-037-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR