4-Chloro-2-fluoro-5-nitrotoluene 98%

Warning

H61841-06 H61841-14 H61841-14 H61841-06
AAH61841-14EA 407.61 USD
AAH61841-14 AAH61841-06 CAAAH61841-06 CAAAH61841-14
4-Chloro-2-fluoro-5-nitrotoluene 98%
4-Chloro-2-fluoro-5-nitrotoluene
Formula: C₇H₅ClFNO₂
Melting Pt: 42…45 °C
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134231
CAS Number: 18349-11-6
EINECS: 242-225-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR