tert-Butyl (5-bromothiazol-2-yl)carbamate 97%

Warning

H61828-06 H61828-03 H61828-06 H61828-03
AAH61828-03EA 101.72 USD
CAAAH61828-03 AAH61828-03 AAH61828-06 CAAAH61828-06
tert-Butyl (5-bromothiazol-2-yl)carbamate 97%
tert-Butyl (5-bromothiazol-2-yl)carbamate
Formula: C8H11BrN2O2S
MW: 279.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368614
CAS Number: 405939-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR