2-Hydroxy-4-(trifluoromethyl)quinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Trifluoromethyl-2(1H)-quinolinone

H61732-MD H61732-03 H61732-03 H61732-MD
AAH61732-MDEA 97.39 USD
CAAAH61732-03 AAH61732-MD AAH61732-03 CAAAH61732-MD
2-Hydroxy-4-(trifluoromethyl)quinoline 97%
2-Hydroxy-4-(trifluoromethyl)quinoline
Formula: C10H6F3NO
MW: 213.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407380
CAS Number: 25199-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR