4-Fluoro-4'-methoxybenzophenone 98%

Warning

Synonyms: (4-Fluorophenyl)(4-methoxyphenyl)methanone

H61679-06 H61679-14 H61679-03 H61679-06 H61679-03
AAH61679-06EA 118.45 USD
AAH61679-06 CAAAH61679-06 AAH61679-14 AAH61679-03 CAAAH61679-03
4-Fluoro-4'-methoxybenzophenone 98%
4-Fluoro-4'-methoxybenzophenone
Formula: C₁₄H₁₁FO₂
MW: 230.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055469
CAS Number: 345-89-1
EINECS: 206-464-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR