2-Chloro-3-methylpyrazine 95%
|
Warning
|
H61651-03
H61651-03
H61651-MD
H61651-MD
AAH61651-MDEA
45.96
USD
CAAAH61651-MD
CAAAH61651-03
AAH61651-MD
AAH61651-03
2-Chloro-3-methylpyrazine 95%
2-Chloro-3-methylpyrazine
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023279 CAS Number: 95-58-9 EINECS: 202-434-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...