3-Chloro-4-methylpyridine 95%

Danger

H61626-06 H61626-14 H61626-06
AAH61626-06EA 66.4 USD
CAAAH61626-06 AAH61626-06 AAH61626-14
3-Chloro-4-methylpyridine 95%
3-Chloro-4-methylpyridine
Formula: C6H6ClN
MW: 127.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114245
CAS Number: 72093-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR