2-Bromo-5-(trifluoromethyl)anisole 98%

Warning

Synonyms: 4-Bromo-3-methoxybenzotrifluoride

H61599-MD H61599-MD H61599-06 H61599-03 H61599-06 H61599-03
AAH61599-06EA 334.11 USD
AAH61599-06 AAH61599-MD CAAAH61599-06 CAAAH61599-03 AAH61599-03 CAAAH61599-MD
2-Bromo-5-(trifluoromethyl)anisole 98%
2-Bromo-5-(trifluoromethyl)anisole
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115974
CAS Number: 402-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR