2,4-Difluorobenzeneboronic acid-1,3-propanediol ester 98%

Synonyms: 2-(2,4-Difluorophenyl)-1,3,2-dioxaborinane

H61525-03 H61525-06 H61525-06 H61525-03
AAH61525-03EA 63.93 USD
CAAAH61525-06 CAAAH61525-03 AAH61525-03 AAH61525-06
2,4-Difluorobenzeneboronic acid-1,3-propanediol ester 98%
2,4-Difluorobenzeneboronic acid-1,3-propanediol ester
Formula: C₉H₉BF₂O₂
MW: 197.976 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504975
CAS Number: 1073372-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR