Anti-DARC Rabbit Polyclonal Antibody

Supplier: ProSci
4071
89416-228EA 530.92 USD
89416-228
Anti-DARC Rabbit Polyclonal Antibody
Antibodies
DARC Antibody: DARC, also known as the Duffy antigen/chemokine receptor, is a seven-transmembrane protein homologous to the classical chemokine G-protein coupled receptors (GPCRs) with the exception of the motif required for G protein coupling. DARC can bind with high affinity several chemokines without transducing any signal, suggesting it may modulate the signals normally induced by these chemokines. Recently, DARC was found to interact with KAI1, a four transmembrane protein recently identified as a tumor metastasis suppressor protein. It is thought that tumor cells dislodged from the primary tumor and expressing KAI1 interact with DARC proteins expressed on vascular cells, transmitting a senescent signal to the tumor cells, while tumor cells that have lost KAI1 expression can proliferate and potentially give rise to metastases. At least three isoforms of DARC are known to exist.

Type: Primary
Antigen: DARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR