Anti-KAPPAB RAS1 Rabbit Polyclonal Antibody

Supplier: ProSci
2493
89415-500EA 531.09 USD
89415-500
Anti-KAPPAB RAS1 Rabbit Polyclonal Antibody
Antibodies
KappaB ras1 Antibody: KappaB ras-1 ( kappa B-ras-1) and kappaB-ras-2 are two small proteins that similar to Ras-like small GTPases that associate with I kappa B (I kappa B), an inhibitor of the transcription factor NF-kappa B. I kappa B exists in two homologous forms, I kappa B-alpha and I kappa B-beta, although I kappa B-beta contains a unique 47-amino acid region within its ankyrin domain. While inactive I kappa B-alpha-NF-kappa B complexes can shuttle in and out of the nucleus, I kappa B-beta-NF-kappa B complexes are retained exclusively in the cytoplasm. It is suggested that kappaB-ras proteins preferentially bind to the I kappa B-beta form through this unique insert within the ankyrin region, thus modulating the cellular location of I kappa B-beta and regulating the rate of degradation of I kappa B-beta. This antibody is specific for kappaB-ras1 and has no cross-reactivity to kappaB-ras2.

Type: Primary
Antigen: NKIRAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR