Anti-CCNO Rabbit Polyclonal Antibody

Supplier: Prosci
3859
89416-070EA 531.09 USD
89416-070
Anti-CCNO Rabbit Polyclonal Antibody
Antibodies
Cyclin O Antibody: Cyclin O, also known as CCNO, has recently been identified as a Cdk1- and Cdk2-activating cyclin specific to apoptosis in lymphoid cells. Cyclin O binds to and activates Cdk2 in response to instrinsic apoptotic stimuli such as glucocorticoids or DNA-damaging agents. Supression of Cyclin O expression by shRNA leads to the inhibition of glucocorticoid and DNA-damage-induced apoptosis due to a failure of apical caspase activation while leaving the CD95 death receptor-mediated apoptosis intact. Note: this gene, which had a previous symbol of UNG2, was erroneously identified as a uracil DNA glycosylase. A later publication identified this gene's product as a cyclin protein family member.

Type: Primary
Antigen: CCNO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR