SCIENCEWARE® Elliptical (Egg-Shaped) Spinbar® Magnetic Stirring Bars, Bel-Art
Spinbar®
F37130-0158 F37130-0001
58949-639CS 1063.12 USD InStock
58949-639 58949-621
SCIENCEWARE® Elliptical (Egg-Shaped) Spinbar® Magnetic Stirring Bars, Bel-Art
Stirrers Magnetic Stirring Bars
These egg-shaped stirring bars are designed especially for use in round-bottom vessels as they spin unrestricted by the vessel contour.
  • Well suited for round-bottom flasks; shape ensures complete mixing
  • Offers minimal contact when used in plastic containers
  • Egg shape provides pivot point without a separate ring or abrupt change in contour
  • Egg-shaped magnets
  • PTFE coating
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.