Would you like to log in again?
SCIENCEWARE® Test Tube Racks, Bel-Art
Supplier: Bel-Art Products
SCIENCEWARE®
H18924-0000 H18923-0000
60917-641EA 34.33 USD InStock
60917-641 60917-638
SCIENCEWARE® Test Tube Racks, Bel-Art
Racks Test Tube Racks
Polyethylene racks are available in two sizes to accommodate tubes, vials, serological tubes, and reagent containers.
  • Durable polyethylene construction
  • Chemically resistant
  • Available in a variety of sizes
  • Compatible with multiple serological tube types
One rack has 12 wells with 20.6 mm diameter (¹³⁄₁₆") holes for 16–20 mm tubes and 5 drying pins. The other has 17 wells with 14 mm diameter (⁹⁄₁₆”) holes and 5 drying pins, and holds serological tubes such as Kahn or Wasserman, or other tubes up to 14 mm diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty