Laboratory Portable Bunsen Burner, Global Market Connections
PRO-2121
89207-496CS 1158.05 USD
89207-496
Laboratory Portable Bunsen Burner, Global Market Connections
Heaters Gas Burners
Portable handheld bunsen burner with stand.
Produces temperatures well over 2500°F for up to to 1.5 hours. Manually controlled flame will not flare in any position. Single-knob gas control features trigger-activated electronic auto ignition and on/off switch. Requires butane fuel cylinder (sold separately).