Ionomycin calcium salt (from Streptomyces conglobatus), Sigma-Aldrich®

407952-5MG 407952-10MG 407952-1MG 407952-25MG 407952-5MG
80056-896EA 517.78 USD
CA80056-892 CA80056-890 CA80056-896 CA80056-894 80056-896
Ionomycin calcium salt (from Streptomyces conglobatus), Sigma-Aldrich®
Ionomycin calcium salt

Useful in cell activation experiments when calcium dose-response data are not required


White to off-white solid appearance. Solubility: DMSO (100 mg/ml).

Formula: C₄₁H₇₀CaO₉
MDL Number: MFCD00151183
CAS Number: 56092-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR