Collagenase, Type I, MilliporeSigma
Supplier: MilliporeSigma
234153-100MG 234153-1GM
80056-218EA 95.57 USD InStock
80056-218 80056-220
Collagenase, Type I, MilliporeSigma
Microbiology Test Systems Microorganism Tests
Crude preparation that contains several collagenases, a sulfhydryl protease, clostripain, and trypsin-like proteolytic activities
Especially useful for tissue dissociation studies. Lyophilized.