Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrocefin ≥95% (by UV method) Dye, Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Warning
Synonyms: 3-(2,4-DINITROSTYRYL)-(6R, 7R)-7-(2-THIENYLACETAMIDO)-CEPH-3-EM-4 CARBOXYLIC ACID, E-ISOMER(6R,7R)-7-[[(E)-4-(2,4-Dinitrophenyl)-2-(2-thienyl)but-3-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acidN-[3-[2-(2,4-DINITRO-PHENYL)-VINYL]-8-OXO-5-THIA-1-AZA-BICYCLO[4.2.0]OCT-7-YL]-2-THIOPHEN-2-YL-ACETAMIDE

Packaged under inert gas.

484400-5MG 484400-5MG
80017-706EA 190.05 USD InStock
80017-706 CA80017-706
Nitrocefin ≥95% (by UV method) Dye, Calbiochem®, Sigma-Aldrich®
Nitrocefin
A chromogenic β-lactamase substrate that undergoes distinctive color change from yellow (λmax = 390 nm at pH 7.0) to red (λmax = 486 nm at pH 7.0) as the amide bond in the β-lactam ring is hydrolyzed by β-lactamase

Nitrocefin is sensitive to hydrolysis by all known lactamases produced by Gram-positive and Gram-negative bacteria. Also useful for the detection of β-lactamase patterns from bacterial cell extracts by isoelectric focusing. Has been used in competitive inhibition studies in developmental work on β-lactamase-resistant antibiotics.


Formula: C₂₁H₁₆N₄O₈S₂
MW: 516.499 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00864896
CAS Number: 41906-86-9
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty