Would you like to log in again?
General-Purpose Series Laboratory Upright Refrigerators, Thermo Scientific
MR12PA-GARE-TS MR49PA-SAEE-TS MR30PA-SAEE-TS MR49PA-SARE-TS MR30SS-GARE-TS MR49SS-GARE-TS MR25PA-GAEE-TS MR30PA-SARE-TS MR30PA-SARE-TS MR12PA-GAEE-TS MR30PA-GARE-TS MR25SS-SARE-TS MR49PA-GAEE-TS MR49SS-SARE-TS MR49SS-SAEE-TS MR45SS-GARE-TS MR30SS-SAEE-TS MR72SS-GARE-TS MR30SS-SARE-TS MR25SS-GAEE-TS MR45SS-GAEE-TS MR49PA-GARE-TS MR38PA-GARE-TS MR25PA-GARE-TS MR25SS-GAEE-TS MR30SS-SARE-TS MR38PA-GAEE-TS MR12PA-GARE-TS MR49PA-SAEE-TS MR30SS-SAEE-TS MR30SS-GAEE-TS MR25SS-GARE-TS MR49PA-GAEE-TS MR12PA-GAEE-TS MR72PA-GAEE-TS MR45PA-GARE-TS MR30SS-GAEE-TS MR45PA-GAEE-TS MR30PA-GAEE-TS MR45PA-GARE-TS MR25SS-SAEE-TS MR38PA-GAEE-TS MR25PA-SAEE-TS MR25SS-GARE-TS MR72SS-GAEE-TS MR72PA-GARE-TS MR49SS-GAEE-TS MR30SS-GARE-TS MR49SS-SARE-TS MR49SS-SAEE-TS MR72PA-GAEE-TS MR72PA-GARE-TS MR25SS-SAEE-TS MR30PA-GARE-TS MR49PA-GARE-TS MR30PA-SAEE-TS MR25SS-SARE-TS MR45SS-GAEE-TS MR25PA-SARE-TS MR49SS-GAEE-TS MR49SS-GARE-TS MR25PA-SARE-TS MR30PA-GAEE-TS MR45PA-GAEE-TS MR45SS-GARE-TS MR25PA-SAEE-TS MR25PA-GAEE-TS MR25PA-GARE-TS MR49PA-SARE-TS MR72SS-GAEE-TS MR38PA-GARE-TS MR72SS-GARE-TS
89208-892EA 9471.77 USD InStock
89208-892 89208-962 CA89208-918 CA89208-964 CA89208-924 CA89208-972 89208-894 CA89208-920 89208-920 CA89208-890 89208-912 CA89208-908 89208-958 89208-980 89208-978 89208-952 89208-930 89208-992 89208-932 89208-902 CA89208-950 CA89208-960 CA89208-936 CA89208-896 CA89208-902 CA89208-932 CA89208-934 CA89208-892 CA89208-962 CA89208-930 89208-922 89208-904 CA89208-958 89208-890 89208-982 CA89208-944 CA89208-922 89208-942 89208-910 89208-944 CA89208-906 89208-934 89208-898 CA89208-904 CA89208-990 CA89208-984 CA89208-970 89208-924 CA89208-980 CA89208-978 CA89208-982 89208-984 89208-906 CA89208-912 89208-960 89208-918 89208-908 89208-950 89208-900 89208-970 89208-972 CA89208-900 CA89208-910 CA89208-942 CA89208-952 CA89208-898 CA89208-894 89208-896 89208-964 89208-990 89208-936 CA89208-992
General-Purpose Series Laboratory Upright Refrigerators, Thermo Scientific
Refrigerators
Thermo Scientific General Purpose (GP) Series Refrigerators offer the dependability and safety required for a variety of daily lab applications
Operation is maintained by a digital temperature controller with intuitive interface, which offers an excellent temperature stability of ±3°C. Units also feature automatic defrost, 5cm (2") diameter stem mount casters, fin and tube condensers, and CFC-free foamed-in polyurethane insulation.
Certifications: UL listed.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: Thermo Scientific provides several service options, such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care packages. To learn more about these services, simply search "Thermo Cold Storage Technical Service" in the VWR.com search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty