Difco™ WL Nutrient Broth, BD
Supplier: BD Life Sciences
247110 247110
90003-400EA 212.93 USD
CA90003-400 90003-400
Difco™ WL Nutrient Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Difco™ WL Nutrient Broth is used for the cultivation of yeasts, molds and bacteria encountered in brewing and industrial fermentation processes.