Large Scoop, Two-Quart, Perfex
Supplier: Perfex
4003W
22940-066EA 41.49 USD
22940-066
Large Scoop, Two-Quart, Perfex
Scoops
High impact resistant polymer scoop (2 qt.) is perfect for a variety of uses: handling food, chemicals, etc., dispensing loose sorbents and other dry materials
Non-Absorbent. Non-Sparking. Chemically resistant. Hygienic.
Easily cleaned and sanitized.
Certifications: USDA and FDA approved materials.