Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "4-trans-4-propylcyclohexyl-phenylboronic-acid"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 146862-02-4 MDL Number: MFCD06658197 Molecular Formula: C15H23BO2 Molecular Weight: 246.16 Form: Crystal Color: White