Pierce™ Prestained Protein Molecular Weight Marker, Thermo Scientific
Supplier: Thermo Scientific
26612 26612
PI26612EA 75.75 USD
PI26612 CAPI26612
Pierce™ Prestained Protein Molecular Weight Marker, Thermo Scientific
This prestained protein MW marker is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents.
Ordering information: Incluses dye-stained proteins in 62.5mM Tris-HCl (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 10mM DTT, 1.5mM NaN3 and 33% glycerol; Component proteins: beta-galactosidase (120kDa), bovine serum albumin (85kDa), ovalbumin (50kDa), cardonic anhydrase (35kDa), beta-lactoglobulin (25kDa), lysozyme (20kDa)