Personna® Double Edge Prep Razor Blades, Accutec Blades
Supplier: Accutec Blades
Personna®
74-0002 74-0002
89237-604CS 344.57 USD InStock
CA11006-856 89237-604
Personna® Double Edge Prep Razor Blades, Accutec Blades
Razors and Blades
Recognized for superior sharpness and quality, Personna® Double Edge Prep Razor Blades are produced using innovative technologies
The cutting edges of each blade are Comfort-Coated® for super smooth shaves. When the standard double-sided blade is used in surgical procedures, the result is a cleaner, smoother, more precise incision with less tissue trauma.
Personna® Double Edge Prep Razor Blades are designed to fit any standard double edge prep razor handle.
The highest quality stainless streel on the market today is carefully selected for optimal charateristics and corrosion-resistance.
Ordering information: Blades are packaged 100 per pack, 10 packs per case.