Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Excellence Plus Level, XPE Series Analytical Balances, METTLER TOLEDO®

Supplier: Mettler Toledo
Excellence Plus XPE
30087770 30133099 30087924 30087635 30131288 30087700 30087653 30087643
High-resolution analytical balances feature a multicolor touch screen that guides the user for secure and ergonomic weighing of small sample sizes.

  • Capacities from 120 to 520 g
  • Readabilities from 0.01 to 0.1 mg
  • Meets all GLP and ISO 9001 Requirements

They also deliver all the statistics needed for Good Laboratory Practice, supporting the highest requirements for safety, efficiency, and ease of compliance.

All models offer quality management features. The innovative StatusLight™ uses color to indicate the status of the balance at a glance. The StaticDetect™ feature warns the user of potential weighing errors caused by static charge. This patented technology takes the worry out of weighing and provides users with a high level of trust in the results. In addition, all models feature the unique SmartGrid™ weighing pan, which is supported at the rear of the weighing chamber, for more efficient weighing. In combination with the optional weighing kit, which includes ErgoClips®, the SmartGrid™ weighing pan becomes an ergonomic weighing system that can be precisely adapted to specific tasks and can help to increase productivity.

The operating menu can be viewed in 11 different languages. The operating software can be upgraded via the Internet. All models will display 12 different weighing units and available applications include statistics, formulation, piece counting, percent weighing, and density. Settings can be password protected. The level and the minimum weight per USP can be monitored by the user.

Certifications: METTLER TOLEDO® provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. UL listed. CSA certified.

Ordering information: The XPE Series Analytical balances are supplied with power supply, two free samples of the SmartPrep single-use antistatic weighing funnel, solid metal pan for placement atop the SmartGrid™, a Static Detect drip tray, and ErgoClip® basket.




 

ORDER

Expand All / Collapse All
Model Weighing Capacity Linearity Pan/Platform Size Readability Stabilization Time Repeatabilty Supplier No. VWR Catalog Number Unit Price Quantity
XPE105 120 g 0.10 mg 7.8 x 7.3 cm (31/16 x 27/8") 0.01 mg 1.5 s 0.03 mg (100 g) 30087635 10025-660 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
XPE105DR 41/120 g 0.15 mg 7.8 x 7.3 cm (31/16 x 27/8") Full Range: 0.1 mg, Fine Range: 0.01 mg 1.5/2.5 s 0.015 mg 30087924 10147-680 Each Retrieving
Product available on GSA Advantage®
XPE205 220 g 0.10 mg 7.8 x 7.3 cm (31/16 x 27/8") 0.01 mg 2.5 s 0.03 mg (200 g) 30087653 10025-662 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
XPE205DR 220 g 0.15 mg 7.8 x 7.3 cm (31/16 x 27/8") 0.1 mg 1.5 s 0.06 mg (200 g) 30087700 10025-664 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
XPE204 220 g 0.2 mg 7.8 x 7.3 cm (31/16 x 27/8") 0.1 mg 1.5 s 0.07 mg (200 g) 30087643 10025-666 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
XPE304 320 g 0.3 mg 7.8 x 7.3 cm (31/16 x 27/8") 0.1 mg 1.5 s 0.08 mg 30131288 10147-682 Each Retrieving
Product available on GSA Advantage®
XPE504 520 g 0.4 mg 7.8 x 7.3 cm (31/16 x 27/8") 0.1 mg 1.5 s 0.12 mg (500 g) 30087770 10025-668 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
XPE504DR 101/520 g 0.5 mg 7.8 x 7.3 cm (31/16 x 27/8") Full Range: 0.1 mg, Fine Range: 0.01 mg 1.5/1.5 s 0.1 mg 30133099 10147-684 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Communication Interface RFID Ready
Dimensions 26.3W x 48.7D x 32.5H cm (10⅜ x 19³⁄₁₆ x 12¾")
Voltage 110/230V, 50/60Hz, 0.8A

ACCESSORIES

Expand All / Collapse All

LabX Express StarterPacs, METTLER TOLEDO®
Description Model Supplier No. VWR Catalog Number Unit Price Quantity
LabX Balance Express StarterPac - 11153120 97044-254 Each Retrieving
LabX Optional License Excellence Balance - 11153220 97044-256 Each Retrieving
LabX Optional License User Management - 11153102 89173-954 Each Retrieving
LabX Optional License Auto Import/Export - 11153105 89173-958 Each Retrieving
Compact Antistatic Kit - 30090337 10067-998 Each Retrieving
ErgoClips® Solution Kit 97034-154 11140251 97034-154 Each Retrieving
LabX Product Database - 11153103 89173-956 Each Retrieving
SmartPrep Funnel - 30061260 10025-440 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: METTLER TOLEDO® provides several service options, including Installation and Qualification, as well as extended warrantys. To learn more about these services please search "METTLER TOLEDO® Installation and Qualification Services" in the VWR.com search field.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty