HiFi HotStart DNA Polymerase, Kapa Biosystems
Supplier: Kapa Biosystems
KK2502 KK2601 KK2602
103568-586EA 681.48 USD
103568-586 103568-582 103568-584
HiFi HotStart DNA Polymerase, Kapa Biosystems
The HiFi DNA Polymerase is a novel, single-enzyme system that exhibits industry-leading performance when compared with other high fidelity polymerases and polymerase blends. KAPA HiFi has been engineered to have an increased affinity for DNA without the need for accessory protein domains
The intrinsic high processivity of KAPA HiFi DNA Polymerase results in significant improvements to yield, sensitivity, speed, target length, and the ability to amplify difficult templates (e.g GC-rich) - standardize your high fidelity PCRs with a single enzyme.