Germinator 500™ Glass Bead Sterilizer, CellPoint Scientific
Supplier: CELL POINT SCIENTIFIC MS
5-1450
101326-488EA 499 USD
101326-488
Germinator 500™ Glass Bead Sterilizer, CellPoint Scientific
Sterilizers Glass Bead Sterilizers
GERMINATOR 500™ is a convenient effective way to quickly decontaminate your micro-dissecting instruments
No need for messy chemicals or open flames in the lab. Small footprint saves bench space. Effective when rapid decontamination of micro-dissecting instruments is needed. The unique stainless steel glass bead bath remains at a constant 500 degrees F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them. Decontaminates within 15 seconds.
Ordering information: The sterilizer uses 1.5 mm glass beads. Refill beads are available when needed.