α-D-Glucose-1-phosphate dipotassium salt hydrate ≥99%
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Glucose-1-(dipotassium phosphate) hydrate
, Cori Ester dipotassium salt hydrate
01451-5G
102573-640EA
564.86
USD
102573-640
α-D-Glucose-1-phosphate dipotassium salt hydrate ≥99%
α-D-Glucose-1-phosphate dipotassium salt hydrate
Formula:
C₆H₁₁K₂O₉P·H₂O MW: 336,315 g/mol (anhy) Storage Temperature: Freezer |
MDL Number:
MFCD00149435 CAS Number: 5996-14-5 |
Specification Test Results
Molecular Formula | C6H11O9PK2 |
Molecular Weight | 336.3 |
MDL Number | MFCD00149435 |
Storage Temperature | Store at 0-8°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...