Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine-2'-monophosphate ≥97%, Powder


00343-5G 00343-250MG 00343-100MG
102615-098EA 977.72 USD InStock
102615-098 102615-094 102615-092
Adenosine-2'-monophosphate ≥97%, Powder
Adenosine-2'-monophosphate


Formula: C₁₀H₁₄N₅O₇P
MW: 347.224 g/mol
CAS Number: 81012-86-4
Order Now
Specification Test Results
Molecular Formula C10H14N5O7P
Molecular Weight 347.2
MDL Number MFCD00022827
Storage Temperature Store at 0-8°C
Color White

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty