Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
VWR® Digitrate® Pro Digital Bottle-Top Burets
182-026 JENC182-025
89403-026EA 1214.57 USD InStock
89403-026 89403-024
VWR® Digitrate® Pro Digital Bottle-Top Burets
Burets
The Digitrate bottle-top buret provides convenient digital bottle-top titration of most reagents.
  • Borosilicate glass
  • LCD display
  • Safety nozzle
  • Chemically resistant
Constructed of borosilicate glass with fluoropolymer resin components
May be easily dismantled for cleaning without the use of tools. Simple removal of electronics for autoclaving. Digitrate fits directly onto 30 mm neck reservoirs with adapters supplied for use on most laboratory bottles. Accuracy: 0.2%. Reproducibility: 0.1%. Each instrument is individually calibrated against National Institute of Standards and Technology traceable equipment.
Amber, low-profile bottle (sold separately) has a side opening which eliminates the need to remove the Digitrate from the bottle during filling. A safety funnel (also sold separately) has an off-set stem for safer filling of the side neck on the bottle.
Digitrate burets feature large LCD display, safety dispense nozzle protector and “zero” reset button for multiple titrations. Simple user calibration procedure. Precision valve mechanism ensures easy priming with no leakage back into the reservoir. Chemically resistant liquid pathway. Visual indication of last calibration. Head rotates 360° for use in any position.
Ordering information: Supplied with 33, 38 and 45 mm bottle neck adapters and a calibration certificate. Also supplied with manufacturer’s three-year warranty. A bottle is not supplied; amber squat bottles with safety side filler necks are sold separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty