Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
DURAC® Plus™ Armored Liquid-In-Glass Thermometers, Organic Liquid Fill, H-B Instrument®
Supplier: VWR International
60171-3311 60171-3411 60171-3511 B60740-0000
10186-266EA 87.83 USD InStock
10186-266 10186-268 10186-270 30617-780
DURAC® Plus™ Armored Liquid-In-Glass Thermometers, Organic Liquid Fill, H-B Instrument®
Thermometers Glass Thermometers General Purpose Glass Thermometers
Open, nickel plated brass armor protects thermometer from accidental breakage during use and storage.
  • Ideal for use in applications where added protection from minor shocks is needed
  • Mercury free; contain blue organic liquid fill against lead-free, white back glass
  • Individually serialized
  • Tested and inspected by our USA quality team
Certifications: Traceable to NIST.
Ordering information: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty