Would you like to log in again?
Scintillation Vials, Borosilicate Glass, without Closures, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
74506-20
66009-260CS 379.74 USD InStock
66009-260
Scintillation Vials, Borosilicate Glass, without Closures, Kimble Chase
Vials Scintillation Vials
Vials are packaged in cellular trays of 100 pieces
Wider opening facilitates introduction of large size samples such as filter discs. Trays are shrink wrapped to preserve cleanliness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty