Buchner Funnel, Table-Type, CoorsTek
Supplier: CoorsTek
60283
30309-223CS 3466.74 USD
30309-223
Buchner Funnel, Table-Type, CoorsTek
Funnels Filtration Funnels Büchner Funnels
One-piece, table-type ceramic filter funnel with side outlet for vacuum filtration
Glazed inside and out, except for the rim. For 240mm filter paper diameter. Tubulature O.D. is approximately 14.5mm at widest point. Autoclavable.